Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3AP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002776.4 | 3 | 1321 | |||
NR_110565.1 | 4 | 1522 | ||||
GenBank mRNA | AK054878.1 | - | 1 | 1708 |