Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002319.2 | 1 | 3553 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB097788.1 | - | 1 | 353 | Chr. 10 - 93958140 - 93958710 [-] on Build GRCh38 |