Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256394.1 | NP_001243323.1 | 3, 4 | 532 | ||
XM_006717169.3 | XP_006717232.1 | 3, 4 | 447 | |||
XM_006717170.3 | XP_006717233.1 | 3, 4 | 315 | |||
XM_006717173.3 | XP_006717236.1 | 3, 4 | 315 | |||
XM_017014877.1 | XP_016870366.1 | 3, 4 | 315 | |||
GenBank mRNA | BC146810.1 | - | 3, 4 | 288 | 8860369 |