Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002451.2 | 1 | 213 | |||
GenBank mRNA | AK024359.1 | - | 1 | 133 | ||
AK303674.1 | - | 1 | 146 | |||
BC040413.1 | - | 1 | 114 | |||
BC098116.1 | - | 1 | 55 | |||
BC098259.1 | - | 1 | 55 | |||
BC099730.1 | - | 1 | 55 | |||
DA501191.1 | - | 1 | 213 |
Gene Symbol: ZNF718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047874.1 | - | 103 |