Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001564.2 | 1 | 5796 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | M97168.3 | - | 1 | 5792 | Chr. X - 73823445 - 73852753 [-] on Build GRCh38 |