Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135998.2 | NP_001129470.1 | 1 | 360 | ||
NM_019056.6 | NP_061929.2 | 1 | 360 | |||
GenBank mRNA | AA287166.1 | - | 1 | 329 | ||
AY359056.1 | - | 1 | 360 | |||
BC010665.1 | - | 1 | 168 | 3870660 |