Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D2Ertd173e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK083486.1 | - | LIT1672 Nespas |
1 | 3409 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB441735.2 | - | 1 | 252 | Chr. 2 - 174276561 - 174277202 [-] on Build GRCm38 |