Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emx2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY117414.1 | - | LIT1111 EMX2OS |
4 | 1564 |