Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vax2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK044313.1 | - | LIT3405 Vax2OS |
2 | 482 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB642242.1 | - | 2 | 482 | Chr. 6 - 83703461 - 83710806 [-] on Build GRCm38 |