Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rian | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AB063319.1 | - | LIT1623 Rian |
1 | 1319 |