Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK020989.1 | - | LIT1001 Ftx |
5 | 521 |