Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF138745.1 | - | LIT1604 Tsix |
4 | 3511 |