Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_036631.2 | 2 | 123 | |||
GenBank mRNA | AF365932.1 | - | LIT1822 Zim3 |
2 | 128 | |
AK007312.2 | - | 409 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707418.1 | - | 3 | 409 | Chr. 7 - 6976302 - 6977420 [-] on Build GRCm38 |