Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nespas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF319945.1 | - | LIT1669 Nespas |
4 | 796 |