Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir155hg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY096003.1 | - | LIT1587 bic |
2 | 377 |