Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx6os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF022079.1 | - | LIT3325 Dlx6as |
1 | 313 |