Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC287004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY035343.1 | - | LIT1742 PINC |
4 | 6221 |