Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC287004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110636.1 | 4 | 6275 | |||
NR_110638.1 | 4 | 2377 | ||||
NR_110640.1 | 5 | 1708 | ||||
NR_110642.1 | 2 | 1110 |