Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AJ412027.1 | - | LIT1925 BCMS |
4, 5 | 450 |