Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | U12897.1 | - | LIT1674 IPW |
3 | 2048 |