Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E47P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120437.1 | 3 | 597 | |||
NR_120439.1 | 2 | 120 | ||||
NR_120440.1 | 2 | 97 | ||||
GenBank mRNA | AA229829.1 | - | 2 | 97 | ||
BC042060.1 | - | 3 | 597 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM906342.1 | - | 609 | Chr. - - [-] on Build |