Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AV714079.1 | - | LIT1011 Jpx |
588 |