Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E62P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF359419.1 | - | LIT2088 PJCG9 pseudogene |
506 |