Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF041081.1 | - | LIT2191 HERC2P3 |
31 | 6014 |