Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400040.1 | - | LIT1999 ST7OT2 |
2, 3 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI262732.1 | - | 2, 3 | 249 | Chr. 7 - 117072424 - 117145558 [-] on Build GRCh38 |