Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400495.1 | - | LIT2223 IC-SNURF-SNRPN 3 prime noncoding transcripts |
5 | 445 | |
AF400496.1 | - | LIT2212 IC-SNURF-SNRPN 3 prime noncoding transcripts |
1 | 12 | ||
AY362865.1 | - | 3 | 290 |