Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_109980.1 | 5 | 1103 | |||
GenBank mRNA | AF400041.1 | - | 4 | 898 | ||
AF400042.1 | - | LIT2001 ST7OT2 |
5 | 1124 |