Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_109980.1 | 1 | 50 | |||
GenBank mRNA | AF400042.1 | - | LIT2001 ST7OT2 |
1 | 71 | |
CB054673.1 | - | 1 | 59 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI262734.1 | - | 1 | 71 | Chr. 7 - 117072070 - 117090781 [-] on Build GRCh38 |