Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927528.1 | - | LIT3536 AY927528 |
1 | 23 |