Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK057701.1 | - | LIT1007 Ftx |
4 | 1934 |