Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927430.1 | - | LIT3466 AY927430 |
2 | 133 |