Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927425.1 | - | LIT3464 AY927425 |
1 | 237 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX872667.1 | - | 1 | 420 | Chr. 22 - 46328923 - 46330079 [+] on Build GRCh38 |