Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927631.1 | - | LIT3615 AY927631 |
1 | 199 |