Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927511.1 | - | LIT3521 AY927511 |
3 | 380 |