Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110841.1 | 1 | 65 | |||
GenBank mRNA | AY927511.1 | - | LIT3521 AY927511 |
1 | 65 | |
AY927512.1 | - | LIT3522 AY927512 |
1 | 65 |