Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LVCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | CB338058.1 | - | LIT2121 GA3824 |
3 | 196 |