Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400043.1 | - | LIT1938 ST7OT2 |
1 | 233 |