Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720874.3 | XP_006720937.1 | 10 | 2192 | ||
GenBank mRNA | BX537511.1 | - | 1 | 646 |