Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039982.1 | 5 | 4060 | |||
GenBank mRNA | AK125018.1 | - | 7 | 4250 | ||
AK127318.1 | - | 6 | 3417 | |||
BC035119.1 | - | 6 | 4127 |