Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP1-AS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036489.1 | 3 | 2202 | |||
GenBank mRNA | AK098180.1 | - | 3 | 2156 |