Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273635.1 | XP_005273692.1 | 11 | 9743 | ||
XM_017013823.1 | XP_016869312.1 | 11 | 9778 | |||
XM_017013824.1 | XP_016869313.1 | 11 | 9595 | |||
GenBank mRNA | BX648950.1 | - | 1 | 459 |