Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 30 | 14670 | ||
NM_001204505.2 | NP_001191434.1 | 29 | 14470 | |||
NM_001204506.2 | NP_001191435.1 | 28 | 14422 | |||
NM_001297429.1 | NP_001284358.1 | 25 | 14058 | |||
NM_152910.5 | NP_690874.2 | 29 | 14633 | |||
NM_178009.4 | NP_821077.1 | 30 | 14764 | |||
NR_123714.1 | 28 | 14524 | ||||
GenBank mRNA | AK055427.1 | - | 1 | 95 | ||
AK090591.1 | - | 1 | 1014 | |||
AK130015.1 | - | 1 | 1159 |