Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 30 | 13607 | ||
NM_001204505.2 | NP_001191434.1 | 29 | 13407 | |||
NM_001204506.2 | NP_001191435.1 | 28 | 13359 | |||
NM_001297429.1 | NP_001284358.1 | 25 | 12995 | |||
NM_152910.5 | NP_690874.2 | 29 | 13570 | |||
NM_178009.4 | NP_821077.1 | 30 | 13701 | |||
NR_123714.1 | 28 | 13461 | ||||
GenBank mRNA | AK130015.1 | - | 1 | 96 |