Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055427.1 | - | 1 | 54 | ||
AK090591.1 | - | 1 | 973 | |||
AK130015.1 | - | 1 | 1118 |