Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717526.1 | XP_006717589.1 | 13 | 2525 | ||
XM_011519755.2 | XP_011518057.1 | 13 | 2504 | |||
XM_011519756.1 | XP_011518058.1 | 12 | 2420 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK091354.1 | - | 1 | 2060 | Chr. 10 - 33194372 - 33196536 [-] on Build GRCh38 |