Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015230.1 | XP_016870719.1 | 16 | 5149 | ||
XM_017015231.1 | XP_016870720.1 | 14 | 4852 | |||
XM_017015237.1 | XP_016870726.1 | 13 | 4685 | |||
GenBank mRNA | AF043897.1 | - | 1 | 1884 |