Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008332.1 | XP_016863821.1 | 7 | 1166 | ||
GenBank mRNA | AK124036.1 | - | 1 | 182 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA959986.1 | - | 1 | 182 | Chr. 4 - 169731153 - 169731705 [-] on Build GRCh38 |