Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303130.1 | NP_001290059.1 | 9 | 2531 | ||
NM_001303131.1 | NP_001290060.1 | 10 | 3018 | |||
NM_174942.2 | NP_777602.1 | 10 | 2652 | |||
GenBank mRNA | BX649059.1 | - | 1 | 1467 |