Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537162.2 | XP_011535464.1 | 10 | 6897 | ||
GenBank mRNA | AL832086.1 | - | 1 | 2738 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA059186.1 | - | 1 | 25 | Chr. 14 - 88591944 - 88592502 [+] on Build GRCh38 |