Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123799.1 | - | 1 | 144 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA681379.1 | - | 1 | 144 | Chr. 7 - 100065038 - 100065603 [-] on Build GRCh38 |